1H-Pyrazolo[4,3-b]pyridine
Supplier: Apollo Scientific
Warning
Synonyms: Pyrazolo[4,3-b]pyridine
OR30695-1G OR30695-250MG OR30695-100MG
APOSOR30695-1GEA 75.4 EUR
APOSOR30695-1G APOSOR30695-250MG APOSOR30695-100MG
1H-Pyrazolo[4,3-b]pyridine
1H-Pyrazolo[4,3-b]pyridine
Formula: C₆H₅N₃
MW: 119.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08234560
CAS Number: 272-52-6