1H-Pyrazolo[4,3-c]pyridine 95%
Supplier: Apollo Scientific
Warning
OR30696-500MG OR30696-250MG
APOSOR30696-500MGEA 40.6 EUR
APOSOR30696-500MG APOSOR30696-250MG
1H-Pyrazolo[4,3-c]pyridine 95%
1H-Pyrazolo[4,3-c]pyridine
Formula: C₆H₅N₃
Storage Temperature: Ambient
MDL Number: MFCD11846327
CAS Number: 271-52-3