3-Bromo-1H-pyrazolo[4,3-c]pyridine
Supplier: Apollo Scientific
Warning
OR30699-1G
APOSOR30699-1GEA 88.6 EUR
APOSOR30699-1G
3-Bromo-1H-pyrazolo[4,3-c]pyridine
3-Bromo-1H-pyrazolo[4,3-c]pyridine
Formula: C₆H₄BrN₃
MW: 198.0200 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11518973
CAS Number: 633328-88-8