(S)-tert-Butyl-3-cyanopyrrolidine-1-carboxylate 97%
Supplier: Apollo Scientific
Synonyms: (S)-1-Boc-3-cyanopyrrolidine, (S)-tert-Butyl 3-cyanopyrrolidine-1-carboxylate
OR307044-1G OR307044-10G OR307044-5G
APOSOR307044-1GEA 102 EUR
APOSOR307044-1G APOSOR307044-10G APOSOR307044-5G
(S)-tert-Butyl-3-cyanopyrrolidine-1-carboxylate 97%
(S)-tert-Butyl-3-cyanopyrrolidine-1-carboxylate
Formula: C₁₀H₁₆N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD03419277
CAS Number: 132945-78-9