(3-Bromobenzyl)methylamine
Supplier: Apollo Scientific
Warning
Synonyms: 1-(3-Bromophenyl)-N-methylmethanamine, 3-Bromo-N-methylbenzylamine
OR307051-100G OR307051-25G OR307051-5G
APOSOR307051-100GEA 219 EUR
APOSOR307051-100G APOSOR307051-25G APOSOR307051-5G
(3-Bromobenzyl)methylamine
(3-Bromobenzyl)methylamine
Formula: C₈H₁₀BrN
Boiling Pt: 231…232 °C
Density: 1.461 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD04507522
CAS Number: 67344-77-8