(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 97+%
Supplier: Apollo Scientific
OR307104-5G
APOSOR307104-5GEA 139 EUR
APOSOR307104-5G
(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid 97+%
(R)-1-(tert-Butoxycarbonyl)pyrrolidine-3-carboxylic acid
Formula: C₁₀H₁₇NO₄
MW: 215.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094727
CAS Number: 72925-16-7