1-Methyl-1H-indazole-5-carboxaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 1-Methyl-1H-indazole-5-carbaldehyde
OR30712-1G OR30712-5G
APOSOR30712-1GEA 65.4 EUR
APOSOR30712-1G APOSOR30712-5G
1-Methyl-1H-indazole-5-carboxaldehyde
1-Methyl-1H-indazole-5-carboxaldehyde
Formula: C₉H₈N₂O
MW: 160.18 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11109365
CAS Number: 872607-89-1