5-Bromo-6-methyl-1H-indazole
Supplier: Apollo Scientific
Warning
OR30742-1G OR30742-5G OR30742-25G
APOSOR30742-1GEA 21.9 EUR
APOSOR30742-1G APOSOR30742-5G APOSOR30742-25G
5-Bromo-6-methyl-1H-indazole
5-Bromo-6-methyl-1H-indazole
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD09026991
CAS Number: 885223-72-3