2-Chloro-6-methoxy-4-methylquinoline
Supplier: Apollo Scientific
OR307810-1G OR307810-5G
APOSOR307810-1GEA 293 EUR
APOSOR307810-1G APOSOR307810-5G
2-Chloro-6-methoxy-4-methylquinoline
2-Chloro-6-methoxy-4-methylquinoline
Formula: C₁₁H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD00087558
CAS Number: 6340-55-2