Ethyl-4-chloro-8-methylquinoline-3-carboxylate
Supplier: Apollo Scientific
Synonyms: 4-Chloro-8-methyl-quinoline-3-carboxylic acid ethyl ester
OR307811-1G OR307811-5G
APOSOR307811-1GEA 71.2 EUR
APOSOR307811-1G APOSOR307811-5G
Ethyl-4-chloro-8-methylquinoline-3-carboxylate
Ethyl 4-chloro-8-methylquinoline-3-carboxylate
Formula: C₁₃H₁₂ClNO₂
MW: 249.70 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00173399
CAS Number: 37041-32-0