6-Bromo-4-methylquinoline
Supplier: Apollo Scientific
Warning
OR307867-1G OR307867-5G
APOSOR307867-1GEA 72.5 EUR
APOSOR307867-1G APOSOR307867-5G
6-Bromo-4-methylquinoline
6-Bromo-4-methylquinoline
Formula: C₁₀H₈BrN
MW: 222.08 g/mol
Melting Pt: 100…101 °C
Storage Temperature: Ambient
MDL Number: MFCD09258893
CAS Number: 41037-28-9