2-Chloro-7-methoxy-3-methylquinoline
Supplier: Apollo Scientific
OR307925-1G OR307925-5G
APOSOR307925-1GEA 301 EUR
APOSOR307925-1G APOSOR307925-5G
2-Chloro-7-methoxy-3-methylquinoline
2-Chloro-7-methoxy-3-methylquinoline
Formula: C₁₁H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD09787682
CAS Number: 132118-45-7