4-Chloro-7-methoxy-2-methylquinoline
Supplier: Apollo Scientific
OR307969-1G OR307969-5G
APOSOR307969-1GEA 310 EUR
APOSOR307969-1G APOSOR307969-5G
4-Chloro-7-methoxy-2-methylquinoline
4-Chloro-7-methoxy-2-methylquinoline
Formula: C₁₁H₁₀ClNO
Storage Temperature: Ambient
MDL Number: MFCD09787735
CAS Number: 75896-68-3