(S)-(+)-1-Aminopropan-2-ol 98%
Supplier: Apollo Scientific
Danger
Synonyms: (S)-1-Aminopropan-2-ol, (S)-(+)-Isopropanolamine, (S)-(+)-1-Amino-2-propanol, (+)-1-Aminopropan-2-ol, (S)-Isopropanolamine, (+)-Isopropanolamine
OR30797-25G OR30797-5G OR30797-1G
APOSOR30797-25GEA 75.4 EUR
APOSOR30797-25G APOSOR30797-5G APOSOR30797-1G
(S)-(+)-1-Aminopropan-2-ol 98%
(S)-(+)-1-Aminopropan-2-ol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 160 °C
Melting Pt: 19…26 °C
Density: 0.954 g/cm³ (25 °C)
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00064429
CAS Number: 2799-17-9
EINECS: 220-533-4
UN: 3259
ADR: 8 II