3-Oxetanamine 97%
Supplier: Apollo Scientific
Danger
Synonyms: 3-amino-oxetane, 3-Aminooxetane
OR308008-1G OR308008-5G OR308008-10G
APOSOR308008-1GEA 46.4 EUR
APOSOR308008-1G APOSOR308008-5G APOSOR308008-10G
3-Oxetanamine 97%
3-Oxetanamine
Formula: C₃H₇NO
MW: 73.09 g/mol
Boiling Pt: 99 °C
Density: 1.05 g/cm³ (25 °C)
Flash Pt: 54 °C
Storage Temperature: Freezer
MDL Number: MFCD08544363
CAS Number: 21635-88-1
UN: 1992
ADR: 3+6.1,III