1-Boc-azetidine-3-ylmethanol 98%
Supplier: Apollo Scientific
Warning
Synonyms: N-BOC-AZETIDINE-3-METHANOL, 1-Boc-Azetidine-3-yl-methanol, Tert-butyl 3-(hydroxymethyl)azetidine-1-carboxylate, 1-BOC-azetidine-3-methanol
OR308030-25G OR308030-5G OR308030-1G
APOSOR308030-25GEA 89.9 EUR
APOSOR308030-25G APOSOR308030-5G APOSOR308030-1G
1-Boc-azetidine-3-ylmethanol 98%
1-Boc-azetidine-3-ylmethanol
Formula: C₉H₁₇NO₃
MW: 187.24 g/mol
Boiling Pt: 268…273 °C
Melting Pt: 55 °C
Density: 1.115 g/cm³ (25 °C)
Flash Pt: 117 °C
Storage Temperature: Ambient
MDL Number: MFCD06656141
CAS Number: 142253-56-3