(S)-1-Boc-Piperazine-2-carboxylic acid
Supplier: Apollo Scientific
Synonyms: (S)-1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid, (2S)-1-(tert-Butoxycarbonyl)piperazine-2-carboxylic acid
OR308062-1G OR308062-5G
APOSOR308062-1GEA 119 EUR
APOSOR308062-1G APOSOR308062-5G
(S)-1-Boc-Piperazine-2-carboxylic acid
(S)-1-Boc-Piperazine-2-carboxylic acid
Formula: C₁₀H₁₈N₂O₄
MW: 230.2600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD02179105
CAS Number: 159532-59-9