(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate
Supplier: Apollo Scientific
Warning
Synonyms: 1-tert-butyl 3-methyl (3S)-piperazine-1,3-dicarboxylate
OR308068-1G OR308068-5G OR308068-10G
APOSOR308068-1GEA 36.4 EUR
APOSOR308068-1G APOSOR308068-5G APOSOR308068-10G
(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate
(S)-1-tert-Butyl 3-methyl piperazine-1,3-dicarboxylate
Formula: C₁₁H₂₀N₂O₄
MW: 244.2875 g/mol
Storage Temperature: Ambient
MDL Number: MFCD04115326
CAS Number: 314741-39-4