Pyrazolo[1,5-a]pyridine-3-carboxylic acid 95%
Supplier: Apollo Scientific
Warning
OR308077-5G
APOSOR308077-5GEA 140 EUR
APOSOR308077-5G
Pyrazolo[1,5-a]pyridine-3-carboxylic acid 95%
Pyrazolo[1,5-a]pyridine-3-carboxylic acid
Formula: C₈H₆N₂O₂
MW: 162.1454 g/mol
Melting Pt: 199…201 °C
Storage Temperature: Ambient
MDL Number: MFCD00229214
CAS Number: 16205-46-2