Ethyl-3-hydroxycyclobutanecarboxylate 95%
Supplier: Apollo Scientific
Warning
Synonyms: Ethyl 3-hydroxycyclobutanecarboxylate
OR308100-1G OR308100-5G OR308100-25G
APOSOR308100-1GEA 43.5 EUR
APOSOR308100-1G APOSOR308100-5G APOSOR308100-25G
Ethyl-3-hydroxycyclobutanecarboxylate 95%
Ethyl-3-hydroxycyclobutanecarboxylate
Formula: C₇H₁₂O₃
MW: 144.17 g/mol
Boiling Pt: 120 °C/10mmHg
Density: 1.0727 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD09758972
CAS Number: 17205-02-6