(S)-Morpholin-3-ylmethanol hydrochloride
Supplier: Apollo Scientific
Warning
OR308126-1G OR308126-250MG
APOSOR308126-1GEA 59.6 EUR
APOSOR308126-1G APOSOR308126-250MG
(S)-Morpholin-3-ylmethanol hydrochloride
(S)-Morpholin-3-ylmethanol hydrochloride
Formula: C₅H₁₂ClNO₂
MW: 153.6073 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12547468
CAS Number: 218594-79-7