Dihydrofuran-3(2H)-one
Supplier: Apollo Scientific
Warning
Synonyms: Oxolan-3-one, Tetrahydrofuran-3-one
OR30821-1G OR30821-250MG OR30821-10G
APOSOR30821-1GEA 29 EUR
APOSOR30821-1G APOSOR30821-250MG APOSOR30821-10G
Dihydrofuran-3(2H)-one
Dihydrofuran-3(2H)-one
Formula: C₄H₆O₂
MW: 86.09 g/mol
Boiling Pt: 139…140 °C
Density: 1.1124 g/cm³ (25 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD07778393
CAS Number: 22929-52-8