1H-Pyrazole-4-carboxylic acid 98%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Pyrazolecarboxylic acid
OR30825-5G OR30825-10G OR30825-25G
APOSOR30825-5GEA 21.9 EUR
APOSOR30825-5G APOSOR30825-10G APOSOR30825-25G
1H-Pyrazole-4-carboxylic acid 98%
1H-Pyrazole-4-carboxylic acid
Formula: C₄H₄N₂O₂
MW: 112.09 g/mol
Melting Pt: 282 °C
Storage Temperature: Ambient
MDL Number: MFCD00011558
CAS Number: 37718-11-9
EINECS: 253-614-5