N,N-Dimethyl-4-nitrosoaniline 98%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Nitroso-N,N-dimethylaniline
OR30877-100G OR30877-25G
APOSOR30877-100GEA 150 EUR
APOSOR30877-100G APOSOR30877-25G
N,N-Dimethyl-4-nitrosoaniline 98%
N,N-Dimethyl-4-nitrosoaniline
Formula: C₈H₁₀N₂O
MW: 150.18 g/mol
Boiling Pt: 259
Melting Pt: 84…87 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002063
CAS Number: 138-89-6
EINECS: 205-343-1
UN: 1369
ADR: 4.2 II
Merck Index: 12,06736