Oxetane-3-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR309073-1G
OR309073-250MG
OR309073-5G
APOSOR309073-1GEA
105
EUR
APOSOR309073-1G
APOSOR309073-250MG
APOSOR309073-5G
Oxetane-3-carboxylic acid
Oxetane-3-carboxylic acid
Formula:
C₄H₆O₃ Boiling Pt: 244 °C Density: 1.374 g/cm³ (25 °C) Flash Pt: 115 °C Storage Temperature: Freezer |
MDL Number:
MFCD08669505 CAS Number: 114012-41-8 |