5-Bromo-2-nitrobenzonitrile
Supplier: Apollo Scientific
Warning
OR30919-5G OR30919-1G OR30919-25G
APOSOR30919-5GEA 24.8 EUR
APOSOR30919-5G APOSOR30919-1G APOSOR30919-25G
5-Bromo-2-nitrobenzonitrile
5-Bromo-2-nitrobenzonitrile
Formula: C₇H₃BrN₂O₂
Melting Pt: 116…118 °C
Storage Temperature: Ambient
MDL Number: MFCD10699674
CAS Number: 89642-50-2