2,4-Diaminopyrimidine 96%
Supplier: Apollo Scientific
Warning
OR30933-1G OR30933-25G OR30933-5G
APOSOR30933-1GEA 21.9 EUR
APOSOR30933-1G APOSOR30933-25G APOSOR30933-5G
2,4-Diaminopyrimidine 96%
2,4-Diaminopyrimidine
Formula: C₄H₆N₄
Melting Pt: 143…147 °C
Storage Temperature: Ambient
MDL Number: MFCD00038023
CAS Number: 156-81-0
EINECS: 205-862-3