Ethyl-4-chloro-6-ethoxyquinoline-3-carboxylate
Supplier: Apollo Scientific
Synonyms: 4-Chloro-6-ethoxyquinoline-3-carboxylic acid ethyl ester
OR309350-1G OR309350-5G
APOSOR309350-1GEA 343 EUR
APOSOR309350-1G APOSOR309350-5G
Ethyl-4-chloro-6-ethoxyquinoline-3-carboxylate
Ethyl 4-chloro-6-ethoxyquinoline-3-carboxylate
Formula: C₁₄H₁₄ClNO₃
Storage Temperature: Ambient
MDL Number: MFCD02166834
CAS Number: 112190-03-1