2-Amino-3-chloropyridine 98%
Supplier: Apollo Scientific
Danger
OR30948-100G OR30948-25G
APOSOR30948-100GEA 95.7 EUR
APOSOR30948-100G APOSOR30948-25G
2-Amino-3-chloropyridine 98%
2-Amino-3-chloropyridine
Formula: C₅H₅ClN₂
Melting Pt: 60…64 °C
Storage Temperature: Ambient
MDL Number: MFCD03541052
CAS Number: 39620-04-7