2,6-Dichloro-4-methylpyridin-3-amine 95%
Supplier: Apollo Scientific
Warning
Synonyms: 3-Amino-2,6-dichloro-4-methylpyridine
OR30949-1G OR30949-5G
APOSOR30949-1GEA 34.8 EUR
APOSOR30949-1G APOSOR30949-5G
2,6-Dichloro-4-methylpyridin-3-amine 95%
2,6-Dichloro-4-methylpyridin-3-amine
Formula: C₆H₆Cl₂N₂
MW: 177.03 g/mol
Melting Pt: 83…85 °C
Storage Temperature: Ambient
MDL Number: MFCD01320814
CAS Number: 129432-25-3