(±)-3-Bromo-1,2-propanediol
Supplier: Apollo Scientific
Danger
Synonyms: (±)-3-Bromo-1,2-dihydroxy propane, 3-Bromopropane-1,2-diol
OR3095-25G OR3095-5G
APOSOR3095-25GEA 357 EUR
APOSOR3095-25G APOSOR3095-5G
(±)-3-Bromo-1,2-propanediol
(±)-3-Bromo-1,2-propanediol
Formula: BrH₂CCH(OH)CH₂OH
Boiling Pt: 72…75 °C/0.2mmHg
Density: 1.77 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004711
CAS Number: 4704-77-2
EINECS: 225-186-2
UN: 3265
ADR: 8,II