5-Amino-2-methoxypyrimidine 95%
Supplier: Apollo Scientific
Warning
OR30959-1G OR30959-250MG OR30959-5G
APOSOR30959-1GEA 63.8 EUR
APOSOR30959-1G APOSOR30959-250MG APOSOR30959-5G
5-Amino-2-methoxypyrimidine 95%
5-Amino-2-methoxypyrimidine
Formula: C₅H₇N₃O
MW: 125.1286 g/mol
Melting Pt: 119…120 °C
Storage Temperature: Ambient
MDL Number: MFCD08234878
CAS Number: 56621-89-7