4-Chlorosalicylaldehyde
Supplier: Apollo Scientific
Warning
Synonyms: 4-Chloro-2-hydroxybenzaldehyde
OR30987-1G OR30987-5G
APOSOR30987-1GEA 21.9 EUR
APOSOR30987-1G APOSOR30987-5G
4-Chlorosalicylaldehyde
4-Chlorosalicylaldehyde
Formula: C₇H₅ClO₂
MW: 156.5664 g/mol
Boiling Pt: 230 °C
Melting Pt: 47…52 °C
Storage Temperature: Ambient
MDL Number: MFCD06252499
CAS Number: 2420-26-0