1H-Indole-2-carbonitrile
Supplier: Apollo Scientific
Warning
Synonyms: Indole-2-carbonitrile
OR310037-1G OR310037-250MG
APOSOR310037-1GEA 148 EUR
APOSOR310037-1G APOSOR310037-250MG
1H-Indole-2-carbonitrile
1H-Indole-2-carbonitrile
Formula: C₉H₆N₂
MW: 142.161 g/mol
Melting Pt: 98…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00187566
CAS Number: 36193-65-4