5-(4-Chlorophenyl)-3-iodopyridin-2-amine
Supplier: Apollo Scientific
|
Warning
|
OR310068-1G
OR310068-500MG
APOSOR310068-1GEA
538
EUR
APOSOR310068-1G
APOSOR310068-500MG
5-(4-Chlorophenyl)-3-iodopyridin-2-amine
5-(4-Chlorophenyl)-3-iodopyridin-2-amine
Formula:
C₁₁H₈ClIN₂ Melting Pt: 136…137 °C Storage Temperature: Ambient |
MDL Number:
MFCD22373657 CAS Number: 1356397-46-0 |