7-Hydroxy-4-(morpholin-4-ylmethyl)-2H-chromen-2-one
Supplier: Apollo Scientific
Warning
OR310110-1G OR310110-250MG
APOSOR310110-1GEA 801 EUR
APOSOR310110-1G APOSOR310110-250MG
7-Hydroxy-4-(morpholin-4-ylmethyl)-2H-chromen-2-one
7-Hydroxy-4-(morpholin-4-ylmethyl)-2H-chromen-2-one
Formula: C₁₄H₁₅NO₄
MW: 261.28 g/mol
Melting Pt: 139…142 °C
Storage Temperature: Ambient
MDL Number: MFCD00868668
CAS Number: 25392-50-1