Methyl (2S)-1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate
Supplier: Apollo Scientific
Warning
OR310146-1G OR310146-500MG
APOSOR310146-1GEA 121 EUR
APOSOR310146-1G APOSOR310146-500MG
Methyl (2S)-1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate
Methyl (2S)-1-(3,3-dimethyl-2-oxopentanoyl)pyrrolidine-2-carboxylate
Formula: C₁₃H₂₁NO₄
Storage Temperature: Refrigerator
MDL Number: MFCD02179273
CAS Number: 186268-77-9