4,4'-Biphenyldicarbonitrile
Supplier: Apollo Scientific
Warning
Synonyms: 4,4'-Dicyanobiphenyl, 4,4'-Dibenzonitrile, [1,1'-Biphenyl]-4,4'-dicarbonitrile, 4,4'-Bibenzonitrile
OR310636-1G OR310636-25G OR310636-5G
APOSOR310636-1GEA 37.7 EUR
APOSOR310636-1G APOSOR310636-25G APOSOR310636-5G
4,4'-Biphenyldicarbonitrile
4,4'-Biphenyldicarbonitrile
Formula: C₁₄H₈N₂
MW: 204.23 g/mol
Melting Pt: 236…240 °C
Storage Temperature: Ambient
MDL Number: MFCD00013805
CAS Number: 1591-30-6