6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Supplier: Apollo Scientific
Warning
OR310932-1G OR310932-500MG
APOSOR310932-1GEA 161 EUR
APOSOR310932-1G APOSOR310932-500MG
6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
6-Methoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formula: C₁₀H₁₄ClNO
MW: 199.68 g/mol
Melting Pt: 233…236 °C
Storage Temperature: Ambient
MDL Number: MFCD01717328
CAS Number: 57196-62-0