2,6-Diazaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester hemioxalate 97%
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 2,6-diazaspiro[3.3]heptane-2-carboxylate oxalate(2:1)
OR312002-1G OR312002-5G
APOSOR312002-1GEA 58 EUR
APOSOR312002-1G APOSOR312002-5G
2,6-Diazaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester hemioxalate 97%
2,6-Diazaspiro[3.3]heptane-2-carboxylic acid tert-butyl ester hemioxalate
Formula: C₂₂H₃₈N₄O₈
MW: 486.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12404932
CAS Number: 1041026-71-4