tert-Butyl-3-(2-hydroxyethyl)azetidine-1-carboxylate
Supplier: Apollo Scientific
Warning
Synonyms: tert-Butyl 3-(2-hydroxyethyl)azetidine-1-carboxylate
OR312044-1G OR312044-250MG
APOSOR312044-1GEA 105 EUR
APOSOR312044-1G APOSOR312044-250MG
tert-Butyl-3-(2-hydroxyethyl)azetidine-1-carboxylate
tert-Butyl-3-(2-hydroxyethyl)azetidine-1-carboxylate
Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD17015912
CAS Number: 152537-03-6