7-Methoxy-1H-pyrrolo[2,3-c]pyridine
Supplier: Apollo Scientific
Warning
Synonyms: 7-Methoxy-6-azaindole
OR3130-1G OR3130-250MG
APOSOR3130-1GEA 270 EUR
APOSOR3130-1G APOSOR3130-250MG
7-Methoxy-1H-pyrrolo[2,3-c]pyridine
7-Methoxy-1H-pyrrolo[2,3-c]pyridine
Formula: C₈H₈N₂O
MW: 148.17 g/mol
Melting Pt: 110…114 °C
Storage Temperature: Ambient
MDL Number: MFCD09864857
CAS Number: 160590-40-9