3-Methylpyrrole
Supplier: Apollo Scientific
Warning
Synonyms: 3-Methyl-1H-pyrrole
OR313041-1G OR313041-5G
APOSOR313041-1GEA 127 EUR
APOSOR313041-1G APOSOR313041-5G
3-Methylpyrrole
3-Methylpyrrole
Formula: C₅H₇N
MW: 81.12 g/mol
Boiling Pt: 144 °C
Melting Pt: –48 °C
Density: 0.93 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00083419
CAS Number: 616-43-3
UN: 1993
ADR: 3,III