(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde 95%
Supplier: Apollo Scientific
Warning
Synonyms: (R)-tert-butyl 4-formyl-2,2-dimethyloxazolidine-3-carboxylate, 3-Oxazolidinecarboxylic acid, 4-forMyl-2,2-diMethyl-, 1,1-diMethylethyl ester, (4R)-, tert-Butyl (R)-(+)-4-formyl-2,2-dimethyl-3-oxazolidinecarboxylate
OR313059-5G
APOSOR313059-5GEA 87 EUR
APOSOR313059-5G
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde 95%
(R)-(+)-3-Boc-2,2-dimethyloxazolidine-4-carboxaldehyde
Formula: C₁₁H₁₉NO₄
MW: 229.276 g/mol
Boiling Pt: 67 °C/0.3mmHg
Density: 1.06 g/cm³ (25 °C)
Flash Pt: 102 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00674064
CAS Number: 95715-87-0