1,2,3,4-Tetrahydro-2,7-naphthyridine hydrochloride
Supplier: Apollo Scientific
Warning
OR313062-1G OR313062-250MG
APOSOR313062-1GEA 317 EUR
APOSOR313062-1G APOSOR313062-250MG
1,2,3,4-Tetrahydro-2,7-naphthyridine hydrochloride
1,2,3,4-Tetrahydro-2,7-naphthyridine hydrochloride
Formula: C₈H₁₁ClN₂
Storage Temperature: Refrigerator
MDL Number: MFCD18253770
CAS Number: 1354940-72-9