3,3-Dimethoxybutan-2-one 98+%
Supplier: Apollo Scientific
Warning
OR3133-1G OR3133-25G OR3133-5G
APOSOR3133-1GEA 21.9 EUR
APOSOR3133-1G APOSOR3133-25G APOSOR3133-5G
3,3-Dimethoxybutan-2-one 98+%
3,3-Dimethoxybutan-2-one
Formula: C₆H₁₂O₃
Boiling Pt: 145…146 °C
Density: 0.987 g/cm³ (25 °C)
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00075266
CAS Number: 21983-72-2
UN: 1224
ADR: 3,III