3-Cyano-4-methylpyridine 97%
Supplier: Apollo Scientific
Warning
Synonyms: 4-Methylpyridine-3-carbonitrile, 3-Cyano-4-picoline, 4-Methylnicotinonitrile
OR3147-1G OR3147-5G OR3147-25G
APOSOR3147-1GEA 21.9 EUR
APOSOR3147-1G APOSOR3147-5G APOSOR3147-25G
3-Cyano-4-methylpyridine 97%
3-Cyano-4-methylpyridine
Formula: C₇H₆N₂
MW: 118.14 g/mol
Boiling Pt: 108…111 °C/20mmHg
Melting Pt: 41…44 °C
Storage Temperature: Ambient
MDL Number: MFCD00234272
CAS Number: 5444-01-9