3-Nitro-4-picoline 96%
Supplier: Apollo Scientific
Danger
Synonyms: 4-Methyl-3-nitropyridine
OR3148-25G
APOSOR3148-25GEA 185 EUR
APOSOR3148-25G
3-Nitro-4-picoline 96%
3-Nitro-4-picoline
Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Boiling Pt: 238 °C
Melting Pt: 24…28 °C
Density: 1.228 g/cm³ (25 °C)
Flash Pt: 108.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00051829
CAS Number: 5832-44-0