(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Supplier: Apollo Scientific
Danger
Synonyms: (-)-2-Azabicyclo[2.2.1]hept-5-en-3-one
OR315002-1G OR315002-5G OR315002-25G
APOSOR315002-1GEA 21.9 EUR
APOSOR315002-1G APOSOR315002-5G APOSOR315002-25G
(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one
(1R,4S)-2-Azabicyclo[2.2.1]hept-5-en-3-one
Formula: C₆H₇NO
MW: 109.1259 g/mol
Melting Pt: 94…97 °C
Storage Temperature: Ambient
MDL Number: MFCD00211274
CAS Number: 79200-56-9