1-Chloroisoquinoline
Supplier: Apollo Scientific
Warning
OR315040-25G OR315040-5G OR315040-100G
APOSOR315040-25GEA 105 EUR
APOSOR315040-25G APOSOR315040-5G APOSOR315040-100G
1-Chloroisoquinoline
1-Chloroisoquinoline
Formula: C₉H₆ClN
MW: 163.61 g/mol
Boiling Pt: 274…275 °C/768mmHg
Melting Pt: 31…36 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00024134
CAS Number: 19493-44-8